Assay Details
| Gene Symbol: LOC4352631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765111.1 | XP_015620597.1 | 10 - 11 | 2026 | 97 | |
| GenBank mRNA | AK073095.1 | - | 10 - 11 | 2027 | 97 | |
| AK106536.1 | - | 10 - 11 | 2026 | 97 | ||
More Information
Set Membership: |
|

LOC4352631
Genomic Map