Assay Details
| Gene Symbol: LOC4352631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765111.1 | XP_015620597.1 | 8 - 9 | 1682 | 80 | |
| GenBank mRNA | AK073095.1 | - | 8 - 9 | 1683 | 80 | |
| AK106536.1 | - | 8 - 9 | 1682 | 80 | ||
More Information
Set Membership: |
|

LOC4352631
Genomic Map