Assay Details
| Gene Symbol: LOC4352631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765111.1 | XP_015620597.1 | 9 - 10 | 1805 | 64 | |
| GenBank mRNA | AK073095.1 | - | 9 - 10 | 1806 | 64 | |
| AK106536.1 | - | 9 - 10 | 1805 | 64 | ||
More Information
Set Membership: |
|

LOC4352631
Genomic Map