Assay Details
Gene Symbol: LOC4352809 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765308.2 | XP_015620794.1 | 5 - 6 | 660 | 139 | |
GenBank mRNA | AK101466.1 | - | 5 - 6 | 661 | 139 | |
AK103368.1 | - | 2 - 3 | 154 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
