Assay Details
Gene Symbol: LOC4350081 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762399.1 | XP_015617885.1 | 12 - 13 | 1955 | 104 | |
XM_015762400.2 | XP_015617886.1 | 12 - 13 | 1945 | 104 | ||
GenBank mRNA | AK062044.1 | - | 7 - 8 | 1163 | 104 | |
AK067129.1 | - | 12 - 13 | 1921 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
