Assay Details
Gene Symbol: LOC4350081 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762399.1 | XP_015617885.1 | 3 - 4 | 717 | 119 | |
XM_015762400.2 | XP_015617886.1 | 3 - 4 | 707 | 119 | ||
GenBank mRNA | AK067129.1 | - | 3 - 4 | 683 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
