Assay Details
Gene Symbol: LOC4350081 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762399.1 | XP_015617885.1 | 9 - 10 | 1639 | 135 | |
XM_015762400.2 | XP_015617886.1 | 9 - 10 | 1629 | 135 | ||
GenBank mRNA | AK062044.1 | - | 4 - 5 | 847 | 135 | |
AK067129.1 | - | 9 - 10 | 1605 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() |
