Assay Details
| Gene Symbol: LOC4350552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761129.2 | 8 - 9 | 1112 | 87 | ||
| GenBank mRNA | AK107085.1 | - | 2 - 3 | 164 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4350552| Gene Symbol: LOC4350552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761129.2 | 8 - 9 | 1112 | 87 | ||
| GenBank mRNA | AK107085.1 | - | 2 - 3 | 164 | 87 | |
Set Membership: |
|