Assay Details
| Gene Symbol: LOC4350552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761129.2 | 9 - 10 | 1214 | 63 | ||
| GenBank mRNA | AK107085.1 | - | 3 - 4 | 266 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4350552| Gene Symbol: LOC4350552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761129.2 | 9 - 10 | 1214 | 63 | ||
| GenBank mRNA | AK107085.1 | - | 3 - 4 | 266 | 63 | |
Set Membership: |
|