Assay Details
Gene Symbol: LOC4350674 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015760138.2 | XP_015615624.1 | 11 - 12 | 1380 | 64 | |
GenBank mRNA | AK072076.1 | - | 11 - 12 | 1291 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
