Assay Details
| Gene Symbol: LOC4350816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762091.2 | XP_015617577.1 | 1 - 2 | 160 | 63 | |
| GenBank mRNA | AK102348.1 | - | 1 - 2 | 77 | 63 | |
| AK288325.1 | - | 3040 | 63 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015762091.2 transcripts.
Set Membership: |
|

LOC4350816
Genomic Map