Assay Details
| Gene Symbol: LOC4350816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762091.2 | XP_015617577.1 | 3 - 4 | 648 | 84 | |
| GenBank mRNA | AK288325.1 | - | 3528 | 84 | ||
More Information
Set Membership: |
|

LOC4350816
Genomic Map