Assay Details
| Gene Symbol: LOC4350816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762091.2 | XP_015617577.1 | 6 - 7 | 1115 | 64 | |
| GenBank mRNA | AK102348.1 | - | 6 - 7 | 1032 | 64 | |
| AK288325.1 | - | 3995 | 64 | |||
More Information
Set Membership: |
|

LOC4350816
Genomic Map