Assay Details
| Gene Symbol: LOC9268461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761126.2 | XP_015616612.1 | 1 | 1437 | 113 | |
| GenBank mRNA | AK069580.1 | - | 1 | 1444 | 113 | |
More Information
Set Membership: |
|

LOC9268461
Genomic Map