Assay Details
| Gene Symbol: LOC4328432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771769.2 | XP_015627255.1 | 1 - 2 | 304 | 67 | |
| GenBank mRNA | AK060914.1 | - | 1 - 2 | 222 | 67 | |
More Information
Set Membership: |
|

LOC4328432
Genomic Map