Assay Details
| Gene Symbol: LOC4328432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771769.2 | XP_015627255.1 | 4 - 5 | 1082 | 101 | |
| XM_015771770.2 | XP_015627256.1 | 4 - 5 | 1083 | 101 | ||
| XM_015771772.2 | XP_015627258.1 | 4 - 5 | 950 | 101 | ||
| GenBank mRNA | AK060914.1 | - | 4 - 5 | 1000 | 101 | |
| AK102265.1 | - | 3 - 4 | 1033 | 101 | ||
More Information
Set Membership: |
|

LOC4328432
Genomic Map