Assay Details
| Gene Symbol: LOC4328515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772111.2 | XP_015627597.1 | 8 - 9 | 1879 | 69 | |
| GenBank mRNA | AK287906.1 | - | 8 - 9 | 1856 | 69 | |
More Information
Set Membership: |
|

LOC4328515
Genomic Map