Assay Details
| Gene Symbol: LOC4328539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772265.2 | XP_015627751.1 | 1 - 2 | 227 | 117 | |
| GenBank mRNA | AK067643.1 | - | 1 - 2 | 224 | 117 | |
More Information
Set Membership: |
|

LOC4328539
Genomic Map