Assay Details
| Gene Symbol: LOC4329388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771860.2 | XP_015627346.1 | 4 - 5 | 909 | 95 | |
| GenBank mRNA | AK073233.1 | - | 4 - 5 | 846 | 95 | |
More Information
Set Membership: |
|

LOC4329388
Genomic Map