Assay Details
| Gene Symbol: LOC4329480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770342.2 | XP_015625828.1 | 1 - 2 | 545 | 86 | |
| GenBank mRNA | AK102498.1 | - | 1 - 2 | 227 | 86 | |
More Information
Set Membership: |
|

LOC4329480
Genomic Map