Assay Details
Gene Symbol: LOC4329643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767619.2 | XP_015623105.1 | 6 - 7 | 736 | 79 | |
XM_015767621.2 | XP_015623107.1 | 6 - 7 | 721 | 79 | ||
GenBank mRNA | AB114856.1 | - | 6 - 7 | 739 | 79 | |
AK100016.1 | - | 3 - 4 | 244 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
