Assay Details
Gene Symbol: LOC4329643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767619.2 | XP_015623105.1 | 8 - 9 | 899 | 85 | |
XM_015767621.2 | XP_015623107.1 | 8 - 9 | 884 | 85 | ||
GenBank mRNA | AB114856.1 | - | 8 - 9 | 902 | 85 | |
AK100016.1 | - | 5 - 6 | 407 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
