Assay Details
| Gene Symbol: LOC4329771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771209.2 | XP_015626695.1 | 1 - 2 | 539 | 66 | |
| GenBank mRNA | AY177696.1 | - | 1 - 2 | 185 | 66 | |
| AY551923.1 | - | 1 - 2 | 335 | 66 | ||
More Information
Set Membership: |
|

LOC4329771
Genomic Map