Assay Details
| Gene Symbol: LOC4329771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771209.2 | XP_015626695.1 | 5 - 6 | 822 | 148 | |
| GenBank mRNA | AY177696.1 | - | 5 - 6 | 468 | 148 | |
More Information
Set Membership: |
|

LOC4329771
Genomic Map