Assay Details
| Gene Symbol: LOC4329771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771209.2 | XP_015626695.1 | 6 - 7 | 909 | 134 | |
| GenBank mRNA | AY177696.1 | - | 6 - 7 | 555 | 134 | |
More Information
Set Membership: |
|

LOC4329771
Genomic Map