Assay Details
| Gene Symbol: LOC4329846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769865.2 | XP_015625351.1 | 4 - 5 | 1974 | 101 | |
| XM_015769866.2 | XP_015625352.1 | 5 - 6 | 1977 | 101 | ||
| GenBank mRNA | AK071429.1 | - | 3 - 4 | 828 | 101 | |
| AK072838.1 | - | 4 - 5 | 1967 | 101 | ||
| AK099686.1 | - | 4 - 5 | 1930 | 101 | ||
More Information
Set Membership: |
|

LOC4329846
Genomic Map