Assay Details
| Gene Symbol: LOC4329846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769865.2 | XP_015625351.1 | 6 - 7 | 2730 | 84 | |
| XM_015769866.2 | XP_015625352.1 | 7 - 8 | 2733 | 84 | ||
| GenBank mRNA | AJ575228.1 | - | 1 - 2 | 523 | 84 | |
| AK060817.1 | - | 5 - 6 | 1188 | 84 | ||
| AK072838.1 | - | 6 - 7 | 2723 | 84 | ||
| AK099686.1 | - | 6 - 7 | 2686 | 84 | ||
| BT129120.1 | - | 1 - 2 | 139 | 84 | ||
| BT130372.1 | - | 1 - 2 | 130 | 84 | ||
More Information
Set Membership: |
|

LOC4329846
Genomic Map