Assay Details
| Gene Symbol: LOC4330076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767164.2 | XP_015622650.1 | 2 - 3 | 690 | 90 | |
| GenBank mRNA | AK058965.1 | - | 2 - 3 | 579 | 90 | |
| AK069855.1 | - | 2 - 3 | 560 | 90 | ||
More Information
Set Membership: |
|

LOC4330076
Genomic Map