Assay Details
| Gene Symbol: LOC4330076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767164.2 | XP_015622650.1 | 3 - 4 | 993 | 64 | |
| GenBank mRNA | AK058965.1 | - | 3 - 4 | 882 | 64 | |
| AK069855.1 | - | 3 - 4 | 863 | 64 | ||
More Information
Set Membership: |
|

LOC4330076
Genomic Map