Assay Details
| Gene Symbol: LOC4330183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771453.2 | XP_015626939.1 | 1 - 2 | 362 | 60 | |
| GenBank mRNA | AK242840.1 | - | 1 - 2 | 199 | 60 | |
More Information
Set Membership: |
|

LOC4330183
Genomic Map