Assay Details
| Gene Symbol: LOC4330183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771453.2 | XP_015626939.1 | 4 - 5 | 701 | 83 | |
| XM_015771454.2 | XP_015626940.1 | 4 - 5 | 775 | 83 | ||
| GenBank mRNA | AK242840.1 | - | 4 - 5 | 538 | 83 | |
More Information
Set Membership: |
|

LOC4330183
Genomic Map