Assay Details
| Gene Symbol: LOC4330183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771453.2 | XP_015626939.1 | 7 - 8 | 932 | 73 | |
| XM_015771454.2 | XP_015626940.1 | 7 - 8 | 1006 | 73 | ||
| GenBank mRNA | AK242840.1 | - | 7 - 8 | 769 | 73 | |
More Information
Set Membership: |
|

LOC4330183
Genomic Map