Assay Details
| Gene Symbol: LOC4330271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771539.2 | XP_015627025.1 | 1 - 2 | 168 | 74 | |
| GenBank mRNA | AB079636.1 | - | 1 - 2 | 160 | 74 | |
| AK242556.1 | - | 1 - 2 | 175 | 74 | ||
More Information
Set Membership: |
|

LOC4330271
Genomic Map