Assay Details
| Gene Symbol: LOC4330479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770349.2 | XP_015625835.1 | 3 - 4 | 1576 | 118 | |
| XM_015770350.2 | XP_015625836.1 | 4 - 5 | 1447 | 118 | ||
| GenBank mRNA | AK106720.1 | - | 3 - 4 | 1473 | 118 | |
More Information
Set Membership: |
|

LOC4330479
Genomic Map