Assay Details
Gene Symbol: LOC4330621 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771308.1 | XP_015626794.1 | 1 - 2 | 526 | 64 | |
XM_015771309.1 | XP_015626795.1 | 1 - 2 | 524 | 64 | ||
GenBank mRNA | AK241644.1 | - | 1 - 2 | 520 | 64 | |
AY177702.1 | - | 1 - 2 | 185 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
