Assay Details
Gene Symbol: LOC4330621 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771308.1 | XP_015626794.1 | 4 - 5 | 761 | 126 | |
XM_015771309.1 | XP_015626795.1 | 4 - 5 | 759 | 126 | ||
GenBank mRNA | AK241644.1 | - | 4 - 5 | 755 | 126 | |
AY177702.1 | - | 4 - 5 | 420 | 126 | ||
AY224482.1 | - | 4 - 5 | 420 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
