Assay Details
Gene Symbol: LOC4330621 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771308.1 | XP_015626794.1 | 6 - 7 | 880 | 73 | |
XM_015771309.1 | XP_015626795.1 | 6 - 7 | 878 | 73 | ||
GenBank mRNA | AK241644.1 | - | 6 - 7 | 874 | 73 | |
AY177702.1 | - | 6 - 7 | 539 | 73 | ||
AY224482.1 | - | 6 - 7 | 539 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
