Assay Details
| Gene Symbol: LOC4330755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767810.1 | XP_015623296.1 | 1 - 2 | 259 | 156 | |
| GenBank mRNA | AK065736.1 | - | 1 - 2 | 174 | 156 | |
| AK104930.1 | - | 1 - 2 | 169 | 156 | ||
More Information
Set Membership: |
|

LOC4330755
Genomic Map