Assay Details
| Gene Symbol: LOC4330812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772119.1 | XP_015627605.1 | 1 - 2 | 452 | 108 | |
| GenBank mRNA | AK059956.1 | - | 1 - 2 | 272 | 108 | |
More Information
Set Membership: |
|

LOC4330812
Genomic Map