Assay Details
| Gene Symbol: LOC4330895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771722.2 | XP_015627208.1 | 2 - 3 | 536 | 67 | |
| GenBank mRNA | AB038319.1 | - | 2 - 3 | 456 | 67 | |
| AK069984.1 | - | 2 - 3 | 537 | 67 | ||
More Information
Set Membership: |
|

LOC4330895
Genomic Map