Assay Details
| Gene Symbol: LOC4330895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771722.2 | XP_015627208.1 | 6 - 7 | 975 | 100 | |
| GenBank mRNA | AK069984.1 | - | 6 - 7 | 976 | 100 | |
More Information
Set Membership: |
|

LOC4330895
Genomic Map