Assay Details
| Gene Symbol: LOC4331064 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771922.2 | XP_015627408.1 | 1 | 1154 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4331064| Gene Symbol: LOC4331064 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771922.2 | XP_015627408.1 | 1 | 1154 | 66 | |
Set Membership: |
|