Assay Details
| Gene Symbol: LOC4331162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769937.2 | XP_015625423.1 | 10 - 11 | 1367 | 72 | |
| XM_015769938.2 | XP_015625424.1 | 11 - 12 | 1274 | 72 | ||
| GenBank mRNA | AK069427.1 | - | 10 - 11 | 1367 | 72 | |
| AK105997.1 | - | 11 - 12 | 1289 | 72 | ||
| AK119352.1 | - | 10 - 11 | 1361 | 72 | ||
More Information
Set Membership: |
|

LOC4331162
Genomic Map