Assay Details
| Gene Symbol: LOC4331162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769937.2 | XP_015625423.1 | 7 - 8 | 1155 | 68 | |
| XM_015769938.2 | XP_015625424.1 | 8 - 9 | 1062 | 68 | ||
| GenBank mRNA | AK058926.1 | - | 2 - 3 | 178 | 68 | |
| AK069427.1 | - | 7 - 8 | 1155 | 68 | ||
| AK105997.1 | - | 8 - 9 | 1062 | 68 | ||
| AK119352.1 | - | 7 - 8 | 1149 | 68 | ||
More Information
Set Membership: |
|

LOC4331162
Genomic Map