Assay Details
| Gene Symbol: LOC4331332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776069.2 | 1 | 701 | 149 | ||
| GenBank mRNA | AK063743.1 | - | 1 | 536 | 149 | |
| D64038.1 | - | 1 | 551 | 149 | ||
More Information
Set Membership: |
|

LOC4331332
Genomic Map