Assay Details
| Gene Symbol: LOC4331378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772592.2 | XP_015628078.1 | 1 - 2 | 1029 | 74 | |
| GenBank mRNA | AK241800.1 | - | 1 - 2 | 724 | 74 | |
More Information
Set Membership: |
|

LOC4331378
Genomic Map