Assay Details
| Gene Symbol: LOC4331415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776147.2 | XP_015631633.1 | 1 - 2 | 297 | 77 | |
| GenBank mRNA | AK073961.1 | - | 1 - 2 | 242 | 77 | |
| AK120855.1 | - | 1 - 2 | 242 | 77 | ||
| X60374.1 | - | 1 - 2 | 146 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the X60374.1, XM_015776147.2 transcripts.
Set Membership: |
|

LOC4331415
Genomic Map