Assay Details
| Gene Symbol: LOC4331415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776147.2 | XP_015631633.1 | 3 - 4 | 596 | 133 | |
| GenBank mRNA | AK120855.1 | - | 3 - 4 | 541 | 133 | |
| X60374.1 | - | 3 - 4 | 445 | 133 | ||
More Information
Set Membership: |
|

LOC4331415
Genomic Map