Assay Details
| Gene Symbol: LOC4331489 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773528.2 | XP_015629014.1 | 1 - 2 | 209 | 119 | |
| GenBank mRNA | AB037900.1 | - | 1 - 2 | 95 | 119 | |
| AK067991.1 | - | 1 - 2 | 184 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015773528.2 transcripts.
Set Membership: |
|

LOC4331489
Genomic Map