Assay Details
| Gene Symbol: LOC4331489 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773528.2 | XP_015629014.1 | 2 - 3 | 343 | 60 | |
| GenBank mRNA | AB037900.1 | - | 2 - 3 | 229 | 60 | |
More Information
Set Membership: |
|

LOC4331489
Genomic Map